Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164627.1 | NP_001158099.1 | 6 - 7 | 510 | 84 | |
NM_001164628.1 | NP_001158100.1 | 5 - 6 | 502 | 84 | ||
NM_001205334.1 | NP_001192263.1 | 6 - 7 | 530 | 84 | ||
NM_028455.4 | NP_082731.2 | 5 - 6 | 522 | 84 | ||
GenBank mRNA | AK014171.1 | - | 5 - 6 | 473 | 84 | |
AK040142.1 | - | 5 - 6 | 472 | 84 | ||
AK086816.1 | - | 5 - 6 | 476 | 84 | ||
AK088242.1 | - | 5 - 6 | 478 | 84 | ||
BC005563.1 | - | 4 - 5 | 465 | 3965919 | 84 | |
BC010306.1 | - | 5 - 6 | 492 | 84 | ||
BY723460.1 | - | 5 - 6 | 496 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |