Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026813.1 | NP_081089.1 | 5 - 6 | 827 | 73 | |
NM_027805.2 | NP_082081.1 | 5 - 6 | 827 | 73 | ||
GenBank mRNA | AK018403.1 | - | 5 - 6 | 747 | 73 | |
AK084684.1 | - | 5 - 6 | 828 | 73 | ||
AK209404.1 | - | 2 - 3 | 268 | 73 | ||
BC024879.1 | - | 4 - 5 | 630 | 4011449 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |