Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026191.2 | NP_080467.3 | 14 - 15 | 1919 | 102 | |
GenBank mRNA | AK004610.1 | - | 14 - 15 | 1883 | 102 | |
AK010512.1 | - | 14 - 15 | 1899 | 102 | ||
AK029935.1 | - | 14 - 15 | 1890 | 102 | ||
AK031000.1 | - | 14 - 15 | 1865 | 102 | ||
AK075991.1 | - | 8 - 9 | 925 | 102 | ||
AK135592.1 | - | 14 - 15 | 1856 | 102 | ||
BC058276.1 | - | 14 - 15 | 1837 | 6515479 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |