Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011815.5 | NP_035945.1 | 11 - 12 | 2114 | 155 | |
GenBank mRNA | AF061744.1 | - | 11 - 12 | 2090 | 155 | |
AK172164.1 | - | 4 - 5 | 346 | 155 | ||
BC141352.1 | - | 11 - 12 | 2055 | 9055964 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |