Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163018.1 | NP_001156490.1 | 4 - 5 | 656 | 116 | |
GenBank mRNA | W13031.1 | - | 2 - 3 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |