Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Omg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019409.2 | NP_062282.2 | 1 - 2 | 180 | 96 | |
GenBank mRNA | AK136910.1 | - | 1 - 2 | 183 | 96 | |
BC024757.1 | - | 1 - 2 | 73 | 5369312 | 96 | |
BY721452.1 | - | 1 - 2 | 180 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |