Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018814.3 | NP_061284.2 | 20 - 21 | 4365 | 120 | |
GenBank mRNA | AF096286.1 | - | 14 - 15 | 2195 | 120 | |
AK129221.1 | - | 15 - 16 | 2245 | 120 | ||
AK154122.1 | - | 1 - 2 | 93 | 120 | ||
AK170320.1 | - | 1 - 2 | 93 | 120 | ||
BC057333.1 | - | 13 - 14 | 2108 | 120 | ||
BC066849.1 | - | 15 - 16 | 1988 | 120 | ||
BC098370.1 | - | 15 - 16 | 1955 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |