Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018814.3 | NP_061284.2 | 23 - 24 | 4800 | 58 | |
GenBank mRNA | AF096286.1 | - | 17 - 18 | 2630 | 58 | |
AK129221.1 | - | 18 - 19 | 2680 | 58 | ||
AK154122.1 | - | 4 - 5 | 528 | 58 | ||
AK170320.1 | - | 4 - 5 | 528 | 58 | ||
BC057333.1 | - | 16 - 17 | 2543 | 58 | ||
BC098370.1 | - | 18 - 19 | 2390 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |