Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00013036.mm.1 |
NM_001177324.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013036.mm.1 |
NM_001177324.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013036.mm.1 |
NM_001253706.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013036.mm.1 |
NM_001253706.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013036.mm.1 |
TC0X00002053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013035.mm.1 |
NM_001177324.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013035.mm.1 |
NM_001177324.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013035.mm.1 |
NM_001253706.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013035.mm.1 |
NM_001253706.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013035.mm.1 |
TC0X00002053.mm.1 |
MTA TC |