Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005507.2 | NP_001005507.1 | 4 - 5 | 413 | 96 | |
NM_001160256.1 | NP_001153728.1 | 4 - 5 | 413 | 96 | ||
NM_001160257.1 | NP_001153729.1 | 4 - 5 | 403 | 96 | ||
GenBank mRNA | AK129101.1 | - | 4 - 5 | 403 | 96 | |
BC082789.1 | - | 4 - 5 | 400 | 30105222 | 96 | |
BC086651.1 | - | 4 - 5 | 413 | 6851591 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |