Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030249.4 | NP_084525.1 | 1 - 2 | 164 | 71 | |
GenBank mRNA | AK029901.1 | - | 1 - 2 | 165 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |