Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243132.1 | NP_001230061.1 | 5 - 6 | 478 | 77 | |
NM_027533.3 | NP_081809.2 | 5 - 6 | 673 | 77 | ||
GenBank mRNA | AK018178.1 | - | 5 - 6 | 476 | 77 | |
AK020982.1 | - | 5 - 6 | 478 | 77 | ||
AK031683.1 | - | 4 - 5 | 382 | 77 | ||
AK032520.1 | - | 5 - 6 | 499 | 77 | ||
AK049888.1 | - | 5 - 6 | 475 | 77 | ||
AK079560.1 | - | 5 - 6 | 477 | 77 | ||
BC007185.1 | - | 5 - 6 | 914 | 3591472 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |