Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164320.1 | NP_001157792.1 | 2 - 3 | 235 | 77 | |
GenBank mRNA | AY100452.1 | - | 2 - 3 | 291 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |