Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172485.3 | NP_766073.2 | 12 - 13 | 2833 | 96 | |
GenBank mRNA | AK051714.1 | - | 12 - 13 | 2675 | 96 | |
AK083798.1 | - | 12 - 13 | 2661 | 96 | ||
BC063250.1 | - | 12 - 13 | 2663 | 6410178 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |