Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162996.1 | NP_001156468.1 | 9 - 10 | 876 | 89 | |
NM_028153.1 | NP_082429.1 | 9 - 10 | 933 | 89 | ||
GenBank mRNA | AK005562.1 | - | 9 - 10 | 899 | 89 | |
AK188717.1 | - | 1 - 2 | 82 | 89 | ||
BC055476.1 | - | 9 - 10 | 876 | 4218133 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |