Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023721.2 | NP_076210.1 | 4 - 5 | 482 | 68 | |
GenBank mRNA | AF298810.1 | - | 4 - 5 | 303 | 68 | |
AK146129.1 | - | 2 - 3 | 153 | 68 | ||
AK146448.1 | - | 4 - 5 | 385 | 68 | ||
AK147910.1 | - | 399 | 68 | |||
AK150981.1 | - | 4 - 5 | 371 | 68 | ||
AK152367.1 | - | 4 - 5 | 371 | 68 | ||
AK153250.1 | - | 4 - 5 | 371 | 68 | ||
AK165761.1 | - | 4 - 5 | 376 | 68 | ||
BC033457.1 | - | 4 - 5 | 344 | 3991107 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |