Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172853.2 | NP_766441.1 | 1 - 2 | 148 | 119 | |
GenBank mRNA | AK034096.1 | - | 1 - 2 | 148 | 119 | |
AK042657.1 | - | 3 - 4 | 543 | 119 | ||
AK140452.1 | - | 4 - 5 | 704 | 119 | ||
BC083189.1 | - | 1 - 2 | 583 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |