Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161767.1 | NP_001155239.1 | 10 - 11 | 1991 | 68 | |
NM_001161768.1 | NP_001155240.1 | 10 - 11 | 1988 | 68 | ||
NM_172451.3 | NP_766039.2 | 10 - 11 | 1987 | 68 | ||
GenBank mRNA | AJ133523.1 | - | 8 - 9 | 1603 | 68 | |
AK028506.1 | - | 10 - 11 | 1987 | 68 | ||
AK049222.1 | - | 10 - 11 | 1990 | 68 | ||
AK155008.1 | - | 10 - 11 | 1994 | 68 | ||
AK159721.1 | - | 10 - 11 | 1969 | 68 | ||
BC119324.1 | - | 8 - 9 | 1640 | 8734073 | 68 | |
BC119326.1 | - | 8 - 9 | 1640 | 8734075 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |