Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008474.2 | NP_032500.2 | 7 - 8 | 1513 | 89 | |
GenBank mRNA | AY028607.1 | - | 7 - 8 | 1470 | 89 | |
BC114971.1 | - | 7 - 8 | 1466 | 40039508 | 89 | |
X65505.1 | - | 4 - 5 | 533 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |