Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029393.1 | NP_083669.1 | 2 - 3 | 730 | 66 | |
GenBank mRNA | AK009986.1 | - | 1 - 2 | 75 | 66 | |
AK053678.1 | - | 2 - 3 | 476 | 66 | ||
BC129847.1 | - | 2 - 3 | 730 | 40107361 | 66 | |
BC129848.1 | - | 2 - 3 | 730 | 40107363 | 66 | |
GU046702.1 | - | 2 - 3 | 699 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |