Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199149.3 | NP_954600.1 | 4 - 5 | 835 | 72 | |
GenBank mRNA | AK050946.1 | - | 4 - 5 | 836 | 72 | |
AK160935.1 | - | 4 - 5 | 835 | 72 | ||
BC070402.1 | - | 4 - 5 | 683 | 5696223 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |