Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285945.1 | NP_001272874.1 | 7 - 8 | 937 | 85 | |
NM_027678.3 | NP_081954.1 | 7 - 8 | 1076 | 85 | ||
GenBank mRNA | AK016921.1 | - | 7 - 8 | 938 | 85 | |
AK042139.1 | - | 7 - 8 | 1044 | 85 | ||
AK050039.1 | - | 3 - 4 | 357 | 85 | ||
AK083641.1 | - | 7 - 8 | 1055 | 85 | ||
AK084943.1 | - | 7 - 8 | 1060 | 85 | ||
BC066035.1 | - | 7 - 8 | 1013 | 6837116 | 85 | |
BC117921.1 | - | 7 - 8 | 891 | 40097463 | 85 | |
BC117922.2 | - | 7 - 8 | 2083 | 40097464 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |