Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025067.1 | NP_001020238.1 | 12 - 13 | 1642 | 68 | |
GenBank mRNA | AK036116.1 | - | 12 - 13 | 1644 | 68 | |
AK155089.1 | - | 12 - 13 | 1705 | 68 | ||
AK173030.1 | - | 2 - 3 | 540 | 68 | ||
BC094228.1 | - | 12 - 13 | 1579 | 30362610 | 68 | |
BC138422.1 | - | 12 - 13 | 1553 | 8861442 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |