Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010269.3 | NP_034399.1 | 11 - 12 | 1511 | 93 | |
NR_104341.1 | 11 - 12 | 1473 | 93 | |||
GenBank mRNA | AK004887.1 | - | 11 - 12 | 1457 | 93 | |
AK082958.1 | - | 11 - 12 | 1473 | 93 | ||
AK146835.1 | - | 11 - 12 | 1501 | 93 | ||
AK161122.1 | - | 11 - 12 | 1410 | 93 | ||
AK166799.1 | - | 11 - 12 | 1426 | 93 | ||
AK166859.1 | - | 11 - 12 | 1425 | 93 | ||
AK188572.1 | - | 2 - 3 | 194 | 93 | ||
AK208171.1 | - | 2 - 3 | 194 | 93 | ||
AK208831.1 | - | 2 - 3 | 194 | 93 | ||
AK213851.1 | - | 2 - 3 | 194 | 93 | ||
BC025070.1 | - | 11 - 12 | 1420 | 3710712 | 93 | |
Y17851.1 | - | 11 - 12 | 1507 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |