Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028111.1 | - | 514 | 140 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 | Galnt6 |
4632410F13, AW047994, GalNAc-T6 |
Anti-sense | Chr. 15 - 100689915 - 100729929 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA I730030J21 gene | I730030J21Rik | 26.3kb | Anti-sense | Chr. 15 - 100730505 - 100732737 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons |