Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163848.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163848.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_001163853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_173186.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
NM_173186.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014272.mm.1 |
TC1700001620.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163847.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163847.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163848.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163848.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163849.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163849.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163850.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_001163853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_173186.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
NM_173186.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014274.mm.1 |
TC1700001620.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163848.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163848.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_001163853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_173186.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
NM_173186.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014273.mm.1 |
TC1700001620.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014276.mm.1 |
NM_001163852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014276.mm.1 |
NM_001163852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014276.mm.1 |
TC1700001620.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014277.mm.1 |
NM_001163852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014277.mm.1 |
NM_001163852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014277.mm.1 |
NM_001163853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014277.mm.1 |
NM_001163853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014277.mm.1 |
TC1700001620.mm.1 |
MTA TC |