Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lima1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113545.1 | NP_001107017.1 | 1 - 2 | 128 | 132 | |
GenBank mRNA | AF307845.1 | - | 1 - 2 | 82 | 132 | |
AK049350.1 | - | 1 - 2 | 78 | 132 | ||
AK078959.1 | - | 1 - 2 | 89 | 132 | ||
CJ073082.1 | - | 1 - 2 | 129 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |