Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700003894.mm.1 |
NM_001286743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003894.mm.1 |
NM_011861.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003894.mm.1 |
NM_178365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003894.mm.1 |
TC1700000464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003887.mm.1 |
NM_001286743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003887.mm.1 |
NM_001286744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003887.mm.1 |
NM_011861.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003887.mm.1 |
TC1700000464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003886.mm.1 |
NM_001286743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003886.mm.1 |
NM_001286744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003886.mm.1 |
NM_011861.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003886.mm.1 |
TC1700000464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003895.mm.1 |
NM_001286743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003895.mm.1 |
NM_001286744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003895.mm.1 |
NM_011861.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003895.mm.1 |
NM_178365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003895.mm.1 |
TC1700000464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003885.mm.1 |
NM_001286743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003885.mm.1 |
NM_001286744.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003885.mm.1 |
NM_011861.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003885.mm.1 |
TC1700000464.mm.1 |
MTA TC |