Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281491.1 | NP_001268420.1 | 6 - 7 | 664 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |