Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134115.2 | NP_598876.1 | 1 - 2 | 229 | 85 | |
GenBank mRNA | AK035731.1 | - | 1 - 2 | 238 | 85 | |
AK080179.1 | - | 1 - 2 | 238 | 85 | ||
AK084433.1 | - | 1 - 2 | 228 | 85 | ||
BC009658.1 | - | 1 - 2 | 229 | 3484764 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |