Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011432.2 | NP_035562.1 | 3 - 4 | 236 | 64 | |
GenBank mRNA | AK137213.1 | - | 3 - 4 | 219 | 64 | |
AK168973.1 | - | 3 - 4 | 209 | 64 | ||
BC008243.1 | - | 3 - 4 | 201 | 2655238 | 64 | |
BC067386.1 | - | 1952 | 64 | |||
BC092266.1 | - | 3 - 4 | 201 | 4009300 | 64 | |
CJ085691.1 | - | 3 - 4 | 64 | |||
U70315.1 | - | 3 - 4 | 205 | 64 | ||
X96767.1 | - | 3 - 4 | 192 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |