Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033218.1 | NP_150087.1 | 11 - 12 | 2309 | 62 | |
GenBank mRNA | AF374267.1 | - | 11 - 12 | 2162 | 62 | |
AK155857.1 | - | 12 - 13 | 2257 | 62 | ||
AK172219.1 | - | 11 - 12 | 2316 | 62 | ||
BC005777.1 | - | 1 - 2 | 51 | 62 | ||
BQ963055.1 | - | 2 - 3 | 364 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |