Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033218.1 | NP_150087.1 | 12 - 13 | 2484 | 71 | |
GenBank mRNA | AF374267.1 | - | 12 - 13 | 2337 | 71 | |
AK155857.1 | - | 13 - 14 | 2432 | 71 | ||
AK172219.1 | - | 12 - 13 | 2491 | 71 | ||
BQ963055.1 | - | 3 - 4 | 539 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |