Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027982.2 | NP_082258.2 | 1 - 2 | 351 | 99 | |
GenBank mRNA | AK009235.1 | - | 1 - 2 | 290 | 99 | |
AY184802.1 | - | 1 - 2 | 351 | 99 | ||
BC117498.1 | - | 1 - 2 | 347 | 40106734 | 99 | |
BC121788.1 | - | 1 - 2 | 346 | 40106732 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |