Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253702.1 | NP_001240631.1 | 4 - 5 | 1110 | 119 | |
NM_001253703.1 | NP_001240632.1 | 4 - 5 | 1110 | 119 | ||
NM_153091.3 | NP_694731.1 | 4 - 5 | 1110 | 119 | ||
NR_045578.1 | 4 - 5 | 1376 | 119 | |||
GenBank mRNA | AB084518.1 | - | 4 - 5 | 524 | 119 | |
AK033865.1 | - | 4 - 5 | 542 | 119 | ||
AK144075.1 | - | 4 - 5 | 550 | 119 | ||
AK148099.1 | - | 4 - 5 | 812 | 119 | ||
AK172102.1 | - | 4 - 5 | 1111 | 119 | ||
AK172218.1 | - | 4 - 5 | 1111 | 119 | ||
AK213571.1 | - | 2 - 3 | 115 | 119 | ||
AK217761.1 | - | 2 - 3 | 115 | 119 | ||
AK219361.1 | - | 2 - 3 | 115 | 119 | ||
BC024473.1 | - | 4 - 5 | 501 | 119 | ||
BC058781.1 | - | 4 - 5 | 535 | 6311899 | 119 | |
BC107394.2 | - | 4 - 5 | 535 | 40058131 | 119 | |
BC145950.1 | - | 4 - 5 | 602 | 40131253 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |