Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008030.1 | NP_032056.1 | 3 - 4 | 359 | 126 | |
GenBank mRNA | AK050123.1 | - | 3 - 4 | 388 | 126 | |
AK149440.1 | - | 3 - 4 | 386 | 126 | ||
U87147.1 | - | 3 - 4 | 359 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |