Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mov10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163440.1 | NP_001156912.1 | 3 - 4 | 594 | 71 | |
NM_001163441.1 | NP_001156913.1 | 3 - 4 | 470 | 71 | ||
NM_008619.2 | NP_032645.2 | 2 - 3 | 571 | 71 | ||
GenBank mRNA | AK004542.1 | - | 3 - 4 | 445 | 71 | |
AK150204.1 | - | 3 - 4 | 454 | 71 | ||
AK159295.1 | - | 3 - 4 | 595 | 71 | ||
AK169206.1 | - | 3 - 4 | 565 | 71 | ||
AK173224.1 | - | 2 - 3 | 484 | 71 | ||
BC053743.1 | - | 2 - 3 | 490 | 6510959 | 71 | |
X52574.1 | - | 2 - 3 | 508 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |