Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033633.3 | NP_001028805.2 | 3 - 4 | 1059 | 68 | |
GenBank mRNA | AK147341.1 | - | 3 - 4 | 1060 | 68 | |
BC119586.1 | - | 4 - 5 | 626 | 40058865 | 68 | |
BC119587.1 | - | 3 - 4 | 568 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |