Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083901.1 | NP_001077370.1 | 7 - 8 | 1003 | 102 | |
NM_133680.3 | NP_598441.3 | 6 - 7 | 1100 | 102 | ||
GenBank mRNA | AK146596.1 | - | 7 - 8 | 996 | 102 | |
BC033469.1 | - | 6 - 7 | 1062 | 4951631 | 102 | |
CN709077.1 | - | 3 - 4 | 429 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |