Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122963.1 | NP_001116435.1 | 4 - 5 | 1514 | 93 | |
NM_028487.4 | NP_082763.3 | 4 - 5 | 1514 | 93 | ||
GenBank mRNA | AK082704.1 | - | 4 - 5 | 1332 | 93 | |
AK135728.1 | - | 4 - 5 | 1445 | 93 | ||
AK136894.1 | - | 4 - 5 | 1293 | 93 | ||
AK145318.1 | - | 4 - 5 | 1503 | 93 | ||
AK151283.1 | - | 3 - 4 | 735 | 93 | ||
AK161928.1 | - | 4 - 5 | 1302 | 93 | ||
AK163370.1 | - | 4 - 5 | 1362 | 93 | ||
AK163390.1 | - | 4 - 5 | 1516 | 93 | ||
AK169406.1 | - | 4 - 5 | 1309 | 93 | ||
AY382529.1 | - | 4 - 5 | 1312 | 93 | ||
BC067075.1 | - | 4 - 5 | 1288 | 30614988 | 93 | |
BC076635.1 | - | 4 - 5 | 1344 | 93 | ||
BC094888.1 | - | 4 - 5 | 1307 | 30628044 | 93 | |
BC096442.1 | - | 4 - 5 | 1331 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |