Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007994.3 | NP_032020.2 | 5 - 6 | 779 | 81 | |
GenBank mRNA | AJ132692.1 | - | 5 - 6 | 706 | 81 | |
AK146946.1 | - | 5 - 6 | 832 | 81 | ||
AK166898.1 | - | 5 - 6 | 780 | 81 | ||
AK166973.1 | - | 5 - 6 | 785 | 81 | ||
AK168374.1 | - | 5 - 6 | 810 | 81 | ||
BC012720.1 | - | 5 - 6 | 4205515 | 81 | ||
D42083.1 | - | 5 - 6 | 725 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |