Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026072.1 | NP_080348.1 | 11 - 12 | 1259 | 63 | |
GenBank mRNA | AK014025.1 | - | 11 - 12 | 1251 | 63 | |
AK075902.1 | - | 11 - 12 | 1237 | 63 | ||
AK077779.1 | - | 11 - 12 | 1202 | 63 | ||
AK166770.1 | - | 11 - 12 | 1259 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |