Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026072.1 | NP_080348.1 | 2 - 3 | 362 | 98 | |
GenBank mRNA | AK014025.1 | - | 2 - 3 | 346 | 98 | |
AK075902.1 | - | 2 - 3 | 340 | 98 | ||
AK077779.1 | - | 2 - 3 | 305 | 98 | ||
AK166770.1 | - | 2 - 3 | 362 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |