Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026072.1 | NP_080348.1 | 4 - 5 | 615 | 78 | |
GenBank mRNA | AK014025.1 | - | 4 - 5 | 598 | 78 | |
AK075902.1 | - | 4 - 5 | 593 | 78 | ||
AK077779.1 | - | 4 - 5 | 558 | 78 | ||
AK166770.1 | - | 4 - 5 | 615 | 78 | ||
BC025437.1 | - | 4 - 5 | 415 | 5025084 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |