Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038010.2 | NP_001033099.1 | 15 - 16 | 2211 | 91 | |
NM_020004.5 | NP_064388.2 | 15 - 16 | 2214 | 91 | ||
GenBank mRNA | AF254441.1 | - | 15 - 16 | 2151 | 91 | |
AK145205.1 | - | 13 - 14 | 2211 | 91 | ||
AK147042.1 | - | 15 - 16 | 2159 | 91 | ||
AK150225.1 | - | 15 - 16 | 2192 | 91 | ||
AK154866.1 | - | 14 - 15 | 2289 | 91 | ||
AK155347.1 | - | 15 - 16 | 2388 | 91 | ||
AK155791.1 | - | 15 - 16 | 2213 | 91 | ||
AK155842.1 | - | 15 - 16 | 2212 | 91 | ||
AK158079.1 | - | 15 - 16 | 2202 | 91 | ||
AK179662.1 | - | 3 - 4 | 264 | 91 | ||
BC011088.1 | - | 15 - 16 | 2121 | 91 | ||
BC051952.1 | - | 13 - 14 | 2435 | 91 | ||
BC060050.1 | - | 13 - 14 | 2435 | 91 | ||
BC063752.1 | - | 15 - 16 | 2196 | 6333090 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |