Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010593.2 | NP_034723.1 | 3 - 4 | 634 | 57 | |
GenBank mRNA | AK145895.1 | - | 3 - 4 | 619 | 57 | |
AK170934.1 | - | 3 - 4 | 607 | 57 | ||
BC040757.1 | - | 3 - 4 | 583 | 4216772 | 57 | |
BC094461.1 | - | 3 - 4 | 587 | 4459882 | 57 | |
M90365.1 | - | 1 - 2 | 99 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |