Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009951.4 | NP_034081.1 | 10 - 11 | 1510 | 125 | |
GenBank mRNA | AF061569.1 | - | 10 - 11 | 1329 | 125 | |
AK013940.1 | - | 10 - 11 | 1477 | 125 | ||
AK044850.1 | - | 10 - 11 | 1415 | 125 | ||
BC051679.1 | - | 10 - 11 | 1493 | 30008106 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |