Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100031825.mm.1 |
NM_001005341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031825.mm.1 |
NM_001005341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031825.mm.1 |
TC1100003462.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031824.mm.1 |
NM_001005341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031824.mm.1 |
NM_001005341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031824.mm.1 |
TC1100003462.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031827.mm.1 |
NM_001005341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031827.mm.1 |
NM_001005341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031827.mm.1 |
TC1100003462.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100031826.mm.1 |
NM_001005341.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031826.mm.1 |
NM_001005341.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100031826.mm.1 |
TC1100003462.mm.1 |
MTA TC |