Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035887.1 | - | 1 - 2 | 62 | 67 | |
BC072599.1 | - | 1 - 2 | 38 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |