Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ylpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178363.3 | NP_848140.3 | 10 - 11 | 5492 | 87 | |
GenBank mRNA | AB033168.1 | - | 10 - 11 | 3165 | 87 | |
AK162250.1 | - | 2 - 3 | 201 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |